Multi-channel pipettes, mechanical, variable volume, SoftGrip™
5 - 50 µl, 0,02, ±0,8 - ±2,0 %, ±0,35 - ±2,0 %, Hnědá, 200 µl
Katalogové číslo HAMI55019-50
Dodavatel: HAMILTON BONADUZ
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
Katalogové číslo HAMI55019-50
Dodavatel: HAMILTON BONADUZ
Přejít na:
Product Details & Documents
Hamilton SoftGrip pipettes are available as 8-channel and 12-channel adjustable volume manual pipettes in two volume ranges covering 5 to 50 µl and 30 to 300 µl.
- Long tip ejector lever and movable lower body can be rotated a full 360º relative to the handle to provide maximum comfort for left- or right-handed use
- Colour-coded plunger buttons and labels for quick volume identification
- Easily activated tip ejector lever releases all the tips in single stroke
- Can be autoclaved without the need for disassembly or recalibration
Please note: Accuracy and precision is guaranteed when used with Hamilton Universal AdvanTip™ precision pipette tips.
Specifikace
- Barevný kód:Hnědá
- Objem:5 - 50 µl
- Increment:0,02
- Přesnost:±0,8 - ±2,0 %
- Správnost:±0,35 - ±2,0 %
- Pro pipetovací špičky:200 µl
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Calibration labels for pipettes
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Pipette calibration key, for simple calibration in the fieldPipette calibration key, for simple calibration in the fieldHAMI3400203Přihlásit se pro dostupnost1 * 1 KS - 239,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Ejector sleeve for SoftGrip™ single channel pipette, 500 - 1000 µlEjector sleeve for SoftGrip™ single channel pipette, 500 - 1000 µlHAMI54756-0102Přihlásit se pro dostupnost1 * 1 KS - 584,00 Kč