Labels, biohazard
10, Biohazard labels, PIC 312-Tri 315-B7541, Polyester, Černá na žluté, 315×273 mm, 1 label/card
116-0744
: Brady
All signs comply with current European legislation.
: The polyester labels have a permanent acrylic adhesive.
- Barva:Černá na žluté
- Popis:Biohazard labels
- Rozměry:315×273 mm
- Materiál:Polyester
- Sekvence:10
- Model:PIC 312-Tri 315-B7541
- Balené:1 label/card