Orders LinkContactUsLinkComponent
Dissection boards, DissecTable™
Board base, Modrá, 481×656×91 mm
Katalogové číslo 720-1686
Dodavatel:   Simport Scientific
Dissection boards, DissecTable™
Katalogové číslo 720-1686
Dodavatel:  Simport Scientific
Dissection boards, DissecTable™
Dissection boards, DissecTable™
Board base, Modrá, 481×656×91 mm
Přejít na:
Price
6 360,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
HDPE / PS. These dissection boards feature a large cutting area on one side. They include dimensional scales in inches and centimeters, a grid and dimensional circles of different sizes. The other side offers two cutting boards half the size with the same dimensional features printed on each one of them.
  • Drain groove is carved all around the edge of the boards in order to contain fluids
  • All corners have rubber feet giving more stability to the working surface
  • Bases are stackable and will not move sideways during dissecting work
Specifikace
  • Barva:
    Modrá
  • Popis:
    Board base
  • Š×H×V:
    481×656×91 mm