Orders LinkContactUsLinkComponent
Dissection boards, DissecTable™
Dissection boards, DissecTable™
Small board with 3 cutting areas, Bílá, 330,2×279,4×12,5 mm
Katalogové číslo 720-1499
Dodavatel:   Simport Scientific
Dissection boards, DissecTable™
Katalogové číslo 720-1499
Dodavatel:  Simport Scientific
Dissection boards, DissecTable™
Dissection boards, DissecTable™
Small board with 3 cutting areas, Bílá, 330,2×279,4×12,5 mm
Přejít na:
Price
3 950,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
HDPE / PS. These dissection boards feature a large cutting area on one side. They include dimensional scales in inches and centimeters, a grid and dimensional circles of different sizes. The other side offers two cutting boards half the size with the same dimensional features printed on each one of them.
  • Drain groove is carved all around the edge of the boards in order to contain fluids
  • All corners have rubber feet giving more stability to the working surface
  • Bases are stackable and will not move sideways during dissecting work
Specifikace
  • Barva:
    Bílá
  • Popis:
    Small board with 3 cutting areas
  • Š×H×V:
    330,2×279,4×12,5 mm