Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
methylcyklohexan 99%, Reagent Grade

methylcyklohexan 99%, Reagent Grade

5096, Glass bottle, 2,5 L
Catalog # M37889-2.5L
Irritant
Health
Flammable
Supplier:   Honeywell Chemicals
methylcyklohexan 99%, Reagent Grade
Catalog # M37889-2.5L
CAS Number:  108-87-2
methylcyklohexan 99%, Reagent Grade
methylcyklohexan 99%, Reagent Grade

5096, Glass bottle, 2,5 L

CAS Number:  108-87-2
Price
2 530,00 Kč
Log in for availability
1 * 2,5 L
Product Details & Documents
  • ≥95% for general laboratory use.
  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19
  • Merck Index:13,06074
  • Boiling Pt:6,7 %(V)
  • Melting Pt:–126 °C
  • Melting Pt:-126 °C
  • Density:0,769 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Flash Pt:-4 °C
  • Storage Temperature:Ambient
  • UN:2296
  • ADR:3,II
  •  :Density: 0,7693 g/cm³ (20 °C)
  • EINECS:203-624-3

Documents

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 98.5 %
  • Vapor Pressure:
    83.2 mmHg ( 37.7 °C)
  • Refractive Index:
    n20/D 1.422 (lit.)
  • Vapor Density:
    3.4 (vs air)
  • Autoignition Temperature:
    545 °F

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    2,5 L
  • Sekvence:
    5096

Specifications