HANKS' BALANCED SALT SOLUTION, MODIFIED, 1 * 500 mL
Catalog # SIALH9394-500ML
Supplier: Merck
Frequently Bought Together
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Roztěrky, tvar L, jednorázové
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Sáčky do homogenizátoru
Supplier: VWR Collection
VWR®, Misky Petriho, PS
Supplier: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Mikrocentrifugační zkumavky Eppendorf® Protein LoBind®
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Sérologická pipeta, standardní
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
Supplier: VWR Collection