Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Screw caps for T501 sample tubes
Screw caps for T501 sample tubes
15, Screw caps for sample tubes with external threads, red
Katalogové číslo SIMPT502R
Dodavatel:   Simport Scientific
Screw caps for T501 sample tubes
Katalogové číslo SIMPT502R
Dodavatel:  Simport Scientific
Screw caps for T501 sample tubes
Screw caps for T501 sample tubes
15, Screw caps for sample tubes with external threads, red
Přejít na:
Price
3 750,00 Kč
Přihlásit se pro dostupnost
1 * 1.000 KS
Product Details & Documents
Caps are available either with or without a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures.
  • Made of polypropylene
  • Available with or without silicone washer
  • Long skirt and super fast thread design for easy seal
  • Autoclavable
As for tubes, these closures are also made of polypropylene, providing the same coefficient of expansion for both, which further enhances the leakproof qualities of the vials at changing temperatures. The cap features a long skirt and a super-fast thread design that allows it to be removed or sealed with a single turn.
Ordering information: Tubes T501 series sold separately.
Specifikace
  • Popis:
    Screw caps for sample tubes with external threads, red
  • Sekvence:
    15
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.