[EN]LOOP MICROLOOP D 5µL PURPLE STERILE/ 1 * 1.000 KS
Catalog # MEDWMW705/20
Supplier: MEDICAL WIRE EQUIPMENT
[EN]LOOP MICROLOOP D 5µL PURPLE STERILE/ 1 * 1.000 KS
Catalog # MEDWMW705/20
Supplier: MEDICAL WIRE EQUIPMENT
Frequently Bought Together
PCR tubes, 0,2 ml, with flat or domed cap, ABgene®
Supplier: Thermo Fisher Scientific
VWR®, Pipety Pasteurovy, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Ubrousky čisticí
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Roztěrky, tvar L, jednorázové
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Forceps
Supplier: VWR Collection
VWR®, Sérologická pipeta, standardní
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
Supplier: VWR Collection