Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
 

Surface protection papers

Bílá, Roll, 500×600 mm
  FIOR0604R00022
 :   Ahlstrom
Surface protection papers
  FIOR0604R00022
 :  Ahlstrom
Surface protection papers
Surface protection papers

Bílá, Roll, 500×600 mm

 
3 410,00 Kč
   
1 * 1 KS
 

The surface paper protects laboratory benches against acids, toxic, corrosive and radioactive products and glass impact.

  • White absorbent paper 100 g/m² with one-side PE coating 25 g/m²
  • Available as sheets or on rolls
  • Grade 604

For use with valuable materials.

 
  • Balené:
    Roll
  • Barva:
    Bílá
  • D׊:
    500×600 mm