Orders LinkContactUsLinkComponent
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
04, 250 ml, 29/32 NS
Katalogové číslo HECH41911018
Flasks, flat bottom with standard ground joint
Katalogové číslo HECH41911018
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
04, 250 ml, 29/32 NS
Přejít na:
Price
690,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    250 ml
  • Sekvence:
    04