Orders LinkContactUsLinkComponent
Cold traps, Type KF
Cold traps, Type KF
03, Cold trap, KF 29-OK-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity
Katalogové číslo KGWA17371
Dodavatel:   KGW
Cold traps, Type KF
Katalogové číslo KGWA17371
Dodavatel:  KGW
Cold traps, Type KF
Cold traps, Type KF
03, Cold trap, KF 29-OK-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity
Price
17 780,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminium
Specifikace
  • Popis:
    Cold trap, KF 29-OK-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity
  • Sekvence:
    03
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Plastic ring for cold trap, type KF
    Plastic ring for cold trap, type KF
    KGWA1733
    02
    Přihlásit se pro dostupnost
    1 * 1 KS - 1 630,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Cooling finger S 29-GL
    Cooling finger S 29-GL
    KGWA1741
    3
    Přihlásit se pro dostupnost
    1 * 1 KS - 7 160,00 Kč