Trypsin Protease, HyClone™
04, Trypsin 2,5% 10X solution, with 25 g porcine trypsin in HBSS, without calcium, magnesium, EDTA or phenol red (liquid), 100 mL
Catalog # HYCLSV30037.01
Supplier: HyClone products (Cytiva)
Trypsin Protease, HyClone™
Catalog # HYCLSV30037.01
Supplier: HyClone products (Cytiva)
CAS Number:
Jump to:
Product Details & Documents
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
- Derived from porcine pancreas
- Gamma irradiated prior to hydration and filling
- Formulated without calcium and magnesium
Specifications
Specifications
- Bal.:100 mL
- Popis:Trypsin 2,5% 10X solution, with 25 g porcine trypsin in HBSS, without calcium, magnesium, EDTA or phenol red (liquid)
- Sekvence:04