Orders LinkContactUsLinkComponent
Trypsin Protease, HyClone™
01, Trypsin, 0,25% 1X, with 2,5 g porcine trypsin in HBSS with 0,2 g/l EDTA, without calcium, magnesium (liquid), 100 ml
Katalogové číslo SH30042.01
Trypsin Protease, HyClone™
Katalogové číslo SH30042.01
Číslo CAS:  
Trypsin Protease, HyClone™
Trypsin Protease, HyClone™
01, Trypsin, 0,25% 1X, with 2,5 g porcine trypsin in HBSS with 0,2 g/l EDTA, without calcium, magnesium (liquid), 100 ml
Přejít na:
Číslo CAS:  
Price
375,00 Kč
Přihlásit se pro dostupnost
1 * 100 mL
Product Details & Documents
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium
Specifikace
Specifikace
  • Bal.:
    100 ml
  • Popis:
    Trypsin, 0,25% 1X, with 2,5 g porcine trypsin in HBSS with 0,2 g/l EDTA, without calcium, magnesium (liquid)
  • Sekvence:
    01
Specifikace