Orders LinkContactUsLinkComponent
Plugs for test tubes
Plugs for test tubes
04, 12 mm, Žlutá
Catalog # 212-0526
Supplier:   BOETTGER PAUL
Plugs for test tubes
Catalog # 212-0526
Supplier:  BOETTGER PAUL
Plugs for test tubes
Plugs for test tubes
04, 12 mm, Žlutá
Price
474,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
  • Not autoclavable
  • Easy handling with a high, ribbed grip
  • Easy to open by tilting 'one-hand operation'
Specifications
  • Barva:
    Žlutá
  • Sekvence:
    04
  • Velikost:
    12 mm