UV/white light transilluminators
Transilluminator, 365/302 nm UV/white light, 8 W, filter size 200×200 mm, LMW-20, 365/302 nm UV/white light
Katalogové číslo 732-4373
Dodavatel: UVP ULTRA VIOLET PRODUCTS
Přejít na:
Product Details & Documents
These UV/white light transilluminators house white light and UV working areas side-by-side. White light can be used for viewing Coomassie Blue stained gels, silver stained protein gels, autoradiographs and microtitre plates. The UV side can be used for viewing ethidium bromide gels.
- Easily switch between light sources using the rocker switch on the front of the unit
- UV side utilises UVG™ long-life filter
- Clear UV blocking cover is hinged and can be height adjusted to provide a suitable viewing angle or to access the filter surface
W×D×H: 486×337×143 mm (height including UV blocking cover)
Weight: 9,6 kg
Specifikace
- Model:LMW-20
- Vlnová délka:365/302 nm UV/white light
- Popis:Transilluminator, 365/302 nm UV/white light, 8 W, filter size 200×200 mm