Orders LinkContactUsLinkComponent
Slide Boxes
Slide Boxes
03, Oranžová, PS, 50, 172×83×31 mm
Catalog # GLWAK50O
Supplier:   GLW
Slide Boxes
Catalog # GLWAK50O
Supplier:  GLW
Slide Boxes
Slide Boxes
03, Oranžová, PS, 50, 172×83×31 mm
Price
143,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
High quality slide storage boxes for microscopy slides.
Used for storing, transporting and freezing in science, industry and biotechnology.
Specifications
  • Počet sklíček:
    50
  • Barva:
    Oranžová
  • Materiál:
    PS
  • Sekvence:
    03
  • Š×H×V:
    172×83×31 mm
Accessories
Product Information
  • Catalog #
    Sekvence
    Popis
    Barva
    Availability
    Price
  • GLWAZK20
    02
    Divider for slides and coverslips of special sizes
    Šedá
    Log in for availability
    1 * 1 KS - 52,50 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Barva
    Availability
    Price
  • GLWAZK15
    01
    Divider for slides and coverslips of special sizes
    Bílá
    Log in for availability
    1 * 1 MJ - 112,00 Kč