Orders LinkContactUsLinkComponent
Certified colour reference standards
Certified colour reference standards
20, Pt-Co 0 mg Pt/l, certified under ISO 9001 quality system, Glass bottle, 500 mL
Katalogové číslo TINT133991
Dodavatel:   Lovibond Tintometer
Certified colour reference standards
Katalogové číslo TINT133991
Dodavatel:  Lovibond Tintometer
Číslo CAS:  
Certified colour reference standards
Certified colour reference standards
20, Pt-Co 0 mg Pt/l, certified under ISO 9001 quality system, Glass bottle, 500 mL
Přejít na:
Číslo CAS:  
Price
2 100,00 Kč
Přihlásit se pro dostupnost
1 * 500 mL
Product Details & Documents
For routine calibration of colorimeters and verification of test data.
Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.
Documents
Specifikace
Specifikace
  • Typ balení:
    Glass bottle
  • Bal.:
    500 mL
  • Popis:
    Pt-Co 0 mg Pt/l, certified under ISO 9001 quality system
  • Sekvence:
    20