Orders LinkContactUsLinkComponent
Pipette tips
10, Standard, 2 - 20 µl, -, Transparent, Eppendorf®, Gilson®, Refill pack
Katalogové číslo KART9745
Dodavatel:   KARTELL
Pipette tips
Katalogové číslo KART9745
Dodavatel:  KARTELL
Pipette tips
Pipette tips
10, Standard, 2 - 20 µl, -, Transparent, Eppendorf®, Gilson®, Refill pack
Přejít na:
Price
1 100,00 Kč
Přihlásit se pro dostupnost
1 * 480 KS
Product Details & Documents
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • Tips in racks come with snap-on lid
  • Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
  • Containers and tips are autoclavable to +121 °C for 20 minutes
Specifikace
  • Balené:
    Refill pack
  • Sterilní:
    -
  • Objem:
    2 - 20 µl
  • Sekvence:
    10
  • Druh špičky:
    Standard
  • Barva špičky:
    Transparent
  • Pro použití s:
    Eppendorf®, Gilson®