Orders LinkContactUsLinkComponent
Plaster dispenser, QuickFix Elastic
Plaster dispenser, QuickFix Elastic
Plaster refill pack with 45 elastic fabric plasters
Katalogové číslo 118-0482
Dodavatel:   Plum Safety
Plaster dispenser, QuickFix Elastic
Katalogové číslo 118-0482
Dodavatel:  Plum Safety
Plaster dispenser, QuickFix Elastic
Plaster dispenser, QuickFix Elastic
Plaster refill pack with 45 elastic fabric plasters
Přejít na:
Price
1 460,00 Kč
Přihlásit se pro dostupnost
1 * 6 KS
Product Details & Documents
Dispenser with elastic finger plasters made from breathable material. The dispenser contains 90 plasters (2 refills) and can be wall-mounted.
  • Elastic textile fabric: breathable and skin friendly
  • Adapts to movements of the skin and is very comfortable to wear
  • Optimum hold even when used in demanding applications
  • Dispenser can be operated with just one hand
  • Easy to refill
  • Very hygienic since each plaster is individually wrapped
Plaster size: 72×25 mm
Informace o dodávce: Dispenser includes 2×45 plaster strips.
Specifikace
  • Popis:
    Plaster refill pack with 45 elastic fabric plasters