Orders LinkContactUsLinkComponent
Cell counters
Benchtop counter, 320×76×56 mm, 9
  HEATHS5360B
 :   HEATHROW
Cell counters
  HEATHS5360B
 :  HEATHROW
Cell counters
Cell counters
Benchtop counter, 320×76×56 mm, 9
16 170,00 Kč
1 * 1 KS
These laboratory cell counters can measure cells such as basophils, eosinophils, myelocytes, juvenile, etc.
  • Each key records up to 999
  • Alarm at one hundred totals
  • Red and white buttons with grey housing
They can measure the number of cells such as Basophils (BASO), Eosinophils (EOSIN), Myelocytes (MYELO), Juvenile (JUVEN), Stab., Seg., Lymph., Mono., and total.
  • Popis:
    Benchtop counter, 320×76×56 mm
  • Uspořádání:
    9