Microarray plates, HTA™ Plate
HTA Plate, 3-D Aldehyde
Katalogové číslo 731-3019
Dodavatel: Greiner Bio-One
Přejít na:
Product Details & Documents
Plastic plates for high-throughput microarrays. With 96 shallow wells and a low well rim, the HTA™ Plate has been optimised for the simultaneous analysis of multiple samples.
- Available with multifunctional, 3-D Amino, 3-D Aldehyde or 3-D Epoxy surface functionality
- Partition of the plate into four individual strips containing 24 wells each gives flexibility, permitting processing of a variable number of samples each day
- Plate dimensions are compatible with automated systems
- Low well rim of only 0,3 mm
- Low autofluorescence
Specifikace
- Popis:HTA Plate, 3-D Aldehyde