Tip refill systems, Axygen®
03, 1 - 1200 µl, -, Transparent, 8 racks, 96 each (stack rack)
Katalogové číslo 732-1402
Dodavatel: Corning
Přejít na:
Product Details & Documents
Transparent PP, autoclavable.
- Easy to load refill racks
- Certified RNase-, DNase- and endotoxin-free
- Universal fit tips have a bevelled reference and graduated marks
Specifikace
- Balené:8 racks, 96 each (stack rack)
- Sterilní:-
- Objem:1 - 1200 µl
- Sekvence:03
- Barva špičky:Transparent