Lab Consumables
12, Soft inoculation loops, 10 μl, sterile, 500×1, individually wrapped
Dodavatel: Copan
Přejít na:
Product Details & Documents
A variety of high-quality lab consumables to meet laboratories everyday needs. We offer a wide range of transfer pipets, loops, spreaders, tubes, caps, and droppers meeting the highest quality standards. Since we produce them with our in-house technology, we can easily customise them upon customer’s request.
Streaking:
Loops and spreaders
Needles, spreaders, and inoculating loops fitting a multitude of different tasks
- Lubricants, oils, and electrostatic charges free
- Certified calibration
- Hexagonal loop shaft for easy handling
- Evans blue dye accuracy certification
Specifikace
- Balené:500×1, individually wrapped
- Popis:Soft inoculation loops, 10 μl, sterile
- Sekvence:12