COPPER STANDARD FOR AAS, TRACECERT TM 1 * 250 mL
Catalog # SUPL38996-250ML
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
Chlorát 1000 mg/l ve vodě standard for ion chromatography
Supplier: VWR Chemicals
Chlorid draselný 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Colour Reference Solutions, Reag.Ph.Eur.
Supplier: REAGECON
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
pH Standard Solutions, IUPAC, Certified
Supplier: Hach