Orders LinkContactUsLinkComponent
Forceps, simple type, TITAN
Forceps, simple type, TITAN
06, Bent, sharp, serrated, 200 mm
Catalog # HAMMHTC085-20
Supplier:   HAMMACHER KARL
Forceps, simple type, TITAN
Catalog # HAMMHTC085-20
Supplier:  HAMMACHER KARL
Forceps, simple type, TITAN
Forceps, simple type, TITAN
06, Bent, sharp, serrated, 200 mm
Price
2 340,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Sharp forceps made from titanium.
  • Straight or bent
  • Simple type
Specifications
  • Type:
    Bent, sharp, serrated
  • Délka:
    200 mm
  • Sekvence:
    06