Orders LinkContactUsLinkComponent
Flame atomisation systems
103, PTFE impact beads for HF digests
  VARI9910053300
 :   VARIAN
Flame atomisation systems
  VARI9910053300
 :  VARIAN
Flame atomisation systems
Flame atomisation systems
103, PTFE impact beads for HF digests
28 275,72 Kč
1 * 1 KS
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
  • Popis:
    PTFE impact beads for HF digests
  • Sekvence:
    103
Accessories
  •  
    Sekvence
    Popis
  • VARI6910009700
    907
    Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    1 * 1 KS
  •  
    Sekvence
    Popis
  • VARI9910025700
    903
    Glass impact beads
    1 * 5 KS - 6 076,62 Kč
  •  
    Sekvence
    Popis
  • VARI9910093400
    905
    Aqueous spray chamber o-ring
    1 * 1 KS - 2 588,19 Kč
  •  
    Sekvence
    Popis
  • VARI9910093500
    906
    Organics o-ring
    1 * 1 KS - 34 863,84 Kč
  •  
    Sekvence
    Popis
  • VARI9910093600
    904
    Fluorinated mixing paddles
    1 * 1 KS - 7 529,28 Kč
  •  
    Sekvence
    Popis
  • VARI9910105800
    908
    Integrated liquid trap (fluorinated float and magnet)
    1 * 1 KS - 12 685,20 Kč
  •  
    Sekvence
    Popis
  • VARI9910093000
    902
    Nebuliser capillary kit
    1 * 1 KS - 22 853,82 Kč
  •  
    Sekvence
    Popis
  • VARI9910093100
    908
    Integrated nebuliser block
    1 * 1 KS - 40 674,48 Kč
  •  
    Sekvence
    Popis
  • VARI190034100
    001
    Flame atomisation system kit
    1 * 1 KS - 86 177,52 Kč