Removable needles
62, 26, 0,46 mm, 0,26 mm, 5 or AS
HAMI7732-03
The benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line. The result is a zero dead volume connection that simplifies priming.
Custom lengths and point styles are available. You can order your custom made needle according to your specification.
* Large hub
** Kel-F hub
Documents
- Ø vněj.:0,46 mm
- Rozchod:26
- Ø vnitř.:0,26 mm
- Typ špičky:5 or AS
- Sekvence:62