Orders LinkContactUsLinkComponent
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Cover for staining dish, - mm
  631-2502
Staining dishes, WHEATON®
  631-2502
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Cover for staining dish, - mm
2 640,00 Kč
1 * 3 KS
Soda-lime glass and stainless steel.
  • Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.
Documents
  • Popis:
    Cover for staining dish
  • Š×H×V:
    - mm