Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
250 ml, 29/32 NS
Catalog # WITG2.649.125
Supplier:   witeg Labortechnik
Separating funnels, Squibb
Catalog # WITG2.649.125
Supplier:  witeg Labortechnik
Separating funnels, Squibb
Separating funnels, Squibb
250 ml, 29/32 NS
Price
2 020,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Separatory funnels, Squibb, made of borosilicate glass 3.3, stopcock with economy PTFE-needle valve.
  • Easy to read graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    250 ml