Orders LinkContactUsLinkComponent
Adapters, reduction, Preciso
Adapters, reduction, Preciso
20, , Redukce, 45/40 NS, , 34/35,
Katalogové číslo WITG0.500.020
Dodavatel:   witeg Labortechnik
Adapters, reduction, Preciso
Katalogové číslo WITG0.500.020
Dodavatel:  witeg Labortechnik
Adapters, reduction, Preciso
Adapters, reduction, Preciso
20, , Redukce, 45/40 NS, , 34/35,
Přejít na:
Price
1 500,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
 : According to DIN 12257.
Specifikace
  • Rozměry zábrusu:
    45/40 NS
  • Zásuvka:
    34/35
  • Type:
    Redukce
  • Sekvence:
    20