Orders LinkContactUsLinkComponent
Cryoboxes
Cryoboxes
01, Přírodní, 133×133×52 mm
Katalogové číslo RATI5220010
Dodavatel:   RATIOLAB
Cryoboxes
Katalogové číslo RATI5220010
Dodavatel:  RATIOLAB
Cryoboxes
Cryoboxes
01, Přírodní, 133×133×52 mm
Přejít na:
Price
675,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
These cryoboxes are made of polypropylene in natural or other colors. They can be used with sample vessels of differing diameters by removing the grid inserts.
  • Robust hinge with safe snap-on lid
  • Stackable for safe transport
  • Drain bores at the bottom for dew liquids
These cryoboxes are particularly practical and flexible due to their removable inserts. Cleaning of the boxes and the inserts is much simpler than cryoboxes with permanently fitted inserts.
The inserts fit all ratiolab® cryo boxes.
Delivery information: Boxes are supplied without inserts, they need to be ordered separately.
Specifikace
  • Š×H×V vnějš.:
    133×133×52 mm
  • Barva:
    Přírodní
  • Sekvence:
    01