Abbe refractometers, NAR
NAR-1T (liquids), 0 − 95 °Bx, 1,30 − 1,70 nD
ATAG1211
: Atago
The NAR Abbe refractometers can meet many of the requirements for routine measurements.
Single wavelengths and standard temperatures can be measured at extended ranges, as well as elevated temperatures, measurements of solid samples (films, plastic sheets).
- Model:NAR-1T (liquids)
- Rozsah:0 − 95 °Bx
- Index lomu:1,30 − 1,70 nD
- Cat. No.:ATAG1211
- Zdroj světla:LED
- Přesnost teplotní sondy:±0,2 °C
- Měření teploty:5...50 °C
- Teplotní sonda:±0,2 °C
Accessories
- SekvencePopis
- SekvencePopis
- SekvencePopis
- SekvencePopis