Dressing jars with lid
102 mm, 130 mm
Katalogové číslo BOCH8621
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
Jar made of stainless steel.
- Stackable
- Lid with handle
- Very good chemical and temperature resistance
Specifikace
- Ø vněj.:102 mm
- Výška:130 mm