Bossheads, three angles
Duraluminium, 9 - 13 mm
Katalogové číslo BOCH5655
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
Three angled bossheads made of duraluminium (aluminium alloy).
- Angle: 90 + 180 + 270°
- Suitable for multilevel and three dimension constructions
Specifikace
- Materiál:Duraluminium
- Pro tyč Ø:9 - 13 mm