VWR® MultiTray®/MultiBox® plus, Pipette Tips
02, 10 µl tips, crystal, graduated, 0,1 - 10 µl, Transparent, MultiTray® 10×96 tips
612-2913
Tips racked in MultiTrays®. Compatible with most micropipettes, except 612-2913 which is compatible with Gilson® and Labmate® micropipettes. MultiBox® plus is an autoclavable PP box with dustproof, hinged and sliding lid, which may be opened completely or partially. MultiBox® plus is suitable for use with all MultiTrays®.
- Tips fit firmly on micropipettor cones, have straight shape for optimum handling, and offer minimum fluid retention
- MultiBox® plus can be repeatedly autoclaved, but should be placed in an adapted container to avoid contact with the autoclave walls (sterilisation conditions: +121 °C, 1,2 bar, max. 30 minutes, with vessels open)
- Up to a maximum of five MultiBox® plus can be stacked
: Tips supplied pre-assembled in MultiTray® systems.
- Balené:MultiTray® 10×96 tips
- Objem:0,1 - 10 µl
- Sekvence:02
- Druh špičky:10 µl tips, crystal, graduated
- Barva špičky:Transparent