Cables for Electrodes
14, Plug cable combination, K 2 S HT (coaxial electrode plug, 2 m coaxial cable, 135 °C, electrode socket (extension cable)), 2 m
Katalogové číslo 662-2208
Dodavatel: SI Analytics
Přejít na:
Product Details & Documents
Documents
Specifikace
- Popis:Plug cable combination, K 2 S HT (coaxial electrode plug, 2 m coaxial cable, 135 °C, electrode socket (extension cable))
- Délka:2 m
- Sekvence:14