Receiving flasks
2000 ml, Low temperature, KS 35/20
Catalog # BUCH040776
Supplier: BUCHI
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
Specifications
- Zásuvka:KS 35/20
- Type:Low temperature
- Kapacita:2000 ml