Orders LinkContactUsLinkComponent
Screw caps, bakelite
Screw caps, bakelite
08, with Butyl/PTFE seal, 48 mm, Černá
Catalog # SPVAP08045
Supplier:   GRAVIS
Screw caps, bakelite
Catalog # SPVAP08045
Supplier:  GRAVIS
Screw caps, bakelite
Screw caps, bakelite
08, with Butyl/PTFE seal, 48 mm, Černá
Price
9 570,00 Kč
Log in for availability
1 * 100 KS
Product Details & Documents
Documents
Specifications
  • Závit:
    48 mm
  • Barva:
    Černá
  • Popis:
    with Butyl/PTFE seal
  • Sekvence:
    08