Orders LinkContactUsLinkComponent
Comparator kits for water testing
Comparator kits for water testing
Hardness* (total as CaCO₃), 20 - 400 mg/l, 100, 1 KIT
Katalogové číslo 705-0068
Dodavatel:   Hach
Comparator kits for water testing
Katalogové číslo 705-0068
Dodavatel:  Hach
Číslo CAS:  
Comparator kits for water testing
Comparator kits for water testing
Hardness* (total as CaCO₃), 20 - 400 mg/l, 100, 1 KIT
Číslo CAS:  
Price
2 090,00 Kč
Přihlásit se pro dostupnost
1 * 1 KIT
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifikace
Specifikace
  • Bal.:
    1 KIT
  • Popis:
    Hardness* (total as CaCO₃)
  • Testů v kitu:
    100
  • Rozsah:
    20 - 400 mg/l
Specifikace
Accessories
Informace o produktu
  • Katalogové číslo
    Bal.
    Dostupnost
    Price
  • Ammonium molybdate reagent for phosphate determinations, 100 ml
    Ammonium molybdate reagent for phosphate determinations, 100 ml
    HACH11032H
    1
    Přihlásit se pro dostupnost
    1 * 1 KS - 865,00 Kč