Quartz control plates
Product Details & Documents
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
- Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
- Available in three optical rotation values
- Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
Certifications: Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
Informace pro objednávání: Sadlles can be ordered in option.
Informace o dodávce: Supplied with a certificate of verification in a plastic storage case.
Specifikace
Specifikace
- Bal.:1
- Popis:Certifikát UKAS pro kalibraci pro křemennou desku při 589 nm
- Sekvence:05