Reference electrodes
Reference electrode, Ag/AgCl reference system, B 1491 HD, +5...+135 °C, Sklo, KCI 3 mol/l, Ceramic, 12×170 mm, Screw plug head
Catalog # SCOI285111504
Supplier: SI Analytics
Jump to:
Product Details & Documents
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Product Literature
Specifications
- Připojení:Screw plug head
- Ø×D:12×170 mm
- Type:B 1491 HD
- Popis:Reference electrode, Ag/AgCl reference system
- Materiál:Sklo
- Diafragma:Ceramic
- Elektrolyt:KCI 3 mol/l
- Tepl.:+5...+135 °C