Adhesion slides
Adhesion slides with 3 square wells and 4 register marks
Katalogové číslo MARI0906000
Dodavatel: MARIENFELD
Přejít na:
Product Details & Documents
Adhesion slides with frosted ends and 90° ground edges.
Specifikace
- Popis:Adhesion slides with 3 square wells and 4 register marks
- Cat. No.:MARI0906000