VWR®, Pipette Tips, Standard
1, 0,5 - 10 µl, -, Transparent, 10 racks, 96 each
Supplier: VWR Collection
Jump to:
Product Details & Documents
These universal tips are compatible with most micropipettes.
- Non sterile, autoclavable
- Smooth inside surface finish for minimum retention of dispensed liquid
Colour: Transparent (0,5 - 10 µl), yellow (2 - 200 µl and 5 - 200 µl), blue (200 - 1000 µl)
Ordering information: Supplied loose or in racks holding 96 tips.
Specifications
- Balené:10 racks, 96 each
- Sterilní:-
- Objem:0,5 - 10 µl
- Sekvence:1
- Barva špičky:Transparent