VWR®, Pipette Tips, Standard
8, 200 - 1000 µl, -, Blue, 10 racks, 96 each
Dodavatel: VWR Collection
Přejít na:
Product Details & Documents
These universal tips are compatible with most micropipettes.
- Non sterile, autoclavable
- Smooth inside surface finish for minimum retention of dispensed liquid
Colour: Transparent (0,5 - 10 µl), yellow (2 - 200 µl and 5 - 200 µl), blue (200 - 1000 µl)
Ordering information: Supplied loose or in racks holding 96 tips.
Specifikace
- Balené:10 racks, 96 each
- Sterilní:-
- Objem:200 - 1000 µl
- Sekvence:8
- Barva špičky:Blue