Microscope slides
003, 90° ground edges, with frosted ends on both sides, half white, 76×26 mm
Katalogové číslo HECH42407010
Dodavatel: GLASWARENFABRIK KARL HECHT
Přejít na:
Product Details & Documents
Pre-cleaned slides with either cut or ground edges.
Specifikace
- Popis:90° ground edges, with frosted ends on both sides, half white
- D׊:76×26 mm
- Sekvence:003