Orders LinkContactUsLinkComponent
RapID™ System
RapID™ System
RapID™ Inoculation Fluid 1 ml, 20
Catalog # OXOIR8325102
RapID™ System
Catalog # OXOIR8325102
CAS Number:  
RapID™ System
RapID™ System
RapID™ Inoculation Fluid 1 ml, 20
CAS Number:  
Price
810,00 Kč
Log in for availability
1 * 20 KS
Product Details & Documents
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
  • Rapid, accurate identification of over 400 medically important organisms
  • Simple, one-step inoculation
  • Four-hour aerobic incubation
  • Easy-to-read colour reactions
Documents
Specifications
Specifications
  • Bal.:
    20
  • Popis:
    RapID™ Inoculation Fluid 1 ml
Specifications