Large insulating dewar flasks, type 131 - 135
01, Large insulating dewar flasks, type 132
Catalog # KGWA1263
Supplier: KGW
Jump to:
Product Details & Documents
Dewar flasks are made of glass and are reliable and economical with a protective casing out of blue-coated metal. These are used in laboratory techniques, medicinal techniques, biotechnology, storing of CO₂ and pressure-less space for coolants.
- With side grips
- Lid made of metal, blue coated with insulating disc
- Lid with over pressure valve
- Lockable lid and roller base
Certifications: Glass material according to ISO 3585 and Dewar flasks according to DIN 12492.
Specifications
- Popis:Large insulating dewar flasks, type 132
- Sekvence:01
- Cat. No.:KGWA1263
- Kapacita:14 L
- Model:132
- Celková výška:635 mm
- Denní vypařování:0,4 L
Accessories
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice
Product Information
- Catalog #SekvencePopisAvailabilityPrice