Electrode Cables
20, S-connector, socket for cable extension, 3 m
Katalogové číslo SCOI285121632
Dodavatel: SI Analytics
Přejít na:
Product Details & Documents
Electrode cables with a range of connectors to fit most pH meters.
Documents
Specifikace
- Popis:S-connector, socket for cable extension
- Délka:3 m
- Sekvence:20