Orders LinkContactUsLinkComponent
Boxes, with Hinged Lid
without label holder, 228×130×60 mm
Katalogové číslo SEMA0348
Dodavatel:   SEMADENI
Boxes, with Hinged Lid
Katalogové číslo SEMA0348
Dodavatel:  SEMADENI
Boxes, with Hinged Lid
Boxes, with Hinged Lid
without label holder, 228×130×60 mm
Přejít na:
Price
337,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
The practical hinged flip-top box container is made from dimensionally stable Polypropylene. The standard colour is grey and the range includes a total of five different sizes. With lid and container made from a single block, snap-on cap and integral hinge, you can package your small item safely and securely.
  • Heavyweight boxes
  • Rigid and robust
  • Stackable
  • PP, grey
Specifikace
  • Type:
    without label holder
  • Š×H×V:
    228×130×60 mm