Orders LinkContactUsLinkComponent
Hoffmann clips
Hoffmann clips
04, 32 mm
Catalog # MURRL141/04
Supplier:   MURRAY S
Hoffmann clips
Catalog # MURRL141/04
Supplier:  MURRAY S
Hoffmann clips
04, 32 mm
Price
444,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Bunsen screw clip, nickel plated brass fitted with protection plate and hinged bottom bar.
  • Spanning width = Width of gate
Specifications
  • Upínací šířka:
    32 mm
  • Sekvence:
    04